n-Hexane

Danger

Synonyms: HexanenHexaneHexyl hydride

AC61018-0040
200057-746EA 249 USD
200057-746
n-Hexane
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94.3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://us.vwr.com/stibo/web/std.lang.all/70/18/737018.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Additional info pesticide residue analysis: < 20 ng/l
Color scale ≤10 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR