Starch, soluble ACS

Synonyms: StarchVitex® iodine indicatorAmylodextrin

AC42449-5000
200004-298EA 63.3 USD
200004-298
Starch, soluble ACS
Starch, soluble
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Melting Pt: 256…258 °C
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Solubility Passes test
Residue after ignition ≤0.4 %
pH 5.0 to 7.0 (2 % solution at 25.0 °C)
Sensitivity Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR