Molybdic acid ACS

Warning

AC42398-0025
103465-960EA 755 USD
103465-960
Molybdic acid ACS
Molybdic acid
Formula: H₂MoO₄
MW: 161.95 g/mol
Density: 3.1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00036287
CAS Number: 7782-91-4
EINECS: 231-970-5
Merck Index: 12,06322

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration ≥85.0 % (Precipitation titraton of molybdate)
Chloride (Cl) ≤0.002 %
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.2 %
AsO4, PO4, SiO4 (as SiO2) ≤0.001 %
Heavy metals (as Pb) ≤0.003 %
Insoluble matter ≤0.01 % (in dilute NH4OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR