Iron(III) chloride hexahydrate 97+% ACS

Danger

Synonyms: Iron trichloride hexahydrateIron (III) chloride 6-hydrateFerric chloride hexahydrateIron trichloride 6-hydrateFerric chloride 6-hydrate

AC42370-5000 AC42370-0025
102777-116EA 247 USD
102777-116 200004-150
Iron(III) chloride hexahydrate 97+% ACS
Iron (III) chloride hexahydrate
Formula: FeCl₃·6H₂O
MW: 270.3 g/mol
Boiling Pt: 280…285 °C
Melting Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8,III
Merck Index: 13,04048
https://us.vwr.com/stibo/web/std.lang.all/05/51/16640551.jpg

Order Now

Specification Test Results

Appearance Yellow to orange to light brown Adhering powder and/or lumps
Titration Iodometric 97.0 to 102.0 %
Nitrate (NO3) ≤100 ppm
Sulfate (SO4) ≤100 ppm
Calcium (Ca) ≤100 ppm
Copper (Cu) ≤30 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤500 ppm
Zinc (Zn) ≤30 ppm
Ferrous iron (Fe 2+) Passes test (limit about 0.002%)
Phosphorous compounds (as PO4) ≤0.01 %
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR