Cadmium(II) chloride hemipentahydrate ACS

Danger

AC42350-1000
102774-982EA 57.6 USD
102774-982
Cadmium(II) chloride hemipentahydrate ACS
Cadmium (II) chloride hemipentahydrate
Formula: CdCl₂·2.5H₂O
MW: 228.35 g/mol
Melting Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00799071
CAS Number: 7790-78-5
EINECS: 233-296-7
UN: 2570
ADR: 6.1,III
Merck Index: 13,01617

Order Now

Specification Test Results

Appearance White Crystals and/or chunks
Titration Complexometric 79.5 to 81.0 % (CdCl2)
Nitrate and Nitrite (as NO3) ≤0.003 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.005 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤0.005 %
Potassium (K) ≤0.02 %
Sodium (Na) ≤0.05 %
Zinc (Zn) ≤0.05 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR