(R)-(-)-2-Amino-3-methyl-1-butanol 98%

Warning

Synonyms: (R)-(-)-2-Amino-3-methylbutanolD(-)-2-Amino-3-methyl-1-butanolD(-)-ValinolD-Valinol

AC42247-0250 AC42247-0050
102777-840EA 686 USD
102777-840 200004-546
(R)-(-)-2-Amino-3-methyl-1-butanol 98%
(R)-(-)-2-Amino-3-methyl-1-butanol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Melting Pt: 32…36 °C
Density: 0.931 g/cm³
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064297
CAS Number: 4276-09-9

Order Now

Specification Test Results

Appearance White to light yellow Crystals or low melting crystalline mass
Infrared spectrum Conforms
Melting point 32 to 36 °C
GC ≥97.5 %
Specific optical rotation -15° to -18° (20 °C, 589 nm) (c=10, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR