Tris(2-methylphenyl)phosphine 99%

Warning

Synonyms: Tris(o-tolyl)phosphineTri-o-tolylphosphine

AC42232-0050 AC42232-0010
200056-618EA 24.2 USD
200056-618 200004-036
Tris(2-methylphenyl)phosphine 99%
Tris(2-methylphenyl)phosphine
Formula: C₂₁H₂₁P
MW: 304.37 g/mol
Melting Pt: 126…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00008514
CAS Number: 6163-58-2
EINECS: 228-193-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 122 to 126 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR