2,4,6-Triphenyl-1,3,5-triazine 99%

Warning

AC42219-0050 AC42219-0250
200034-316EA 319 USD
200034-316 200064-000
2,4,6-Triphenyl-1,3,5-triazine 99%
2,4,6-Triphenyl-1,3,5-triazine
Formula: C₂₁H₁₅N₃
MW: 309.37 g/mol
Boiling Pt: 542 °C (1013 hPa)
Melting Pt: 231…236 °C
MDL Number: MFCD00006051
CAS Number: 493-77-6
EINECS: 207-779-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 231 to 236 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR