Tricyclohexylphosphine 97%

Warning

AC42161-0050
200021-038EA 71.2 USD
200021-038
Tricyclohexylphosphine 97%
Tricyclohexylphosphine
Formula: C₁₈H₃₃P
MW: 280.43 g/mol
Melting Pt: 77…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00003853
CAS Number: 2622-14-2
EINECS: 220-069-2

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 83 °C
NMR (31P-NMR) ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR