n-Tricosane 98%

Synonyms: Tricosane

AC42160-0250 AC42160-0050 AC42160-0010
103512-542EA 22.6 USD
103512-542 200021-036 103512-580
n-Tricosane 98%
n-Tricosane
Formula: H₃C(CH₂)₂₁CH₃
MW: 324.63 g/mol
Boiling Pt: 380 °C (760 mmHg)
Melting Pt: 47…50 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009350
CAS Number: 638-67-5
EINECS: 211-347-4

Order Now

Specification Test Results

Appearance White Crystals, flakes and/or chunks
Infrared spectrum Conforms
Melting point 47 to 50 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR