Tributyl citrate 99+%

Synonyms: Citric acid tributyl esterHydroxytricarballylic acid tributyl ester

AC42144-2500
103466-676EA 87.4 USD
103466-676
Tributyl citrate 99+%
Tributyl citrate
Formula: C₁₈H₃₂O₇
MW: 360.45 g/mol
Boiling Pt: 325 °C (760 mmHg)
Melting Pt: –20 °C
Density: 1.042 g/cm³
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00027217
CAS Number: 77-94-1
EINECS: 201-071-2
Merck Index: 13,01563

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20 °C, 589 nm)
Water ≤0.3 % (K.F.)
GC ≥99.0 %
Color scale ≤50 APHA
Acidity ≤0.2 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR