2,2,2-Tribromoethanol 99%

Warning

AC42143-0100 AC42143-0500 AC42143-2500
200004-018EA 58.1 USD
200004-018 200004-020 103512-540
2,2,2-Tribromoethanol 99%
2,2,2-Tribromoethanol
Formula: Br₃CCH₂OH
MW: 282.76 g/mol
Melting Pt: 73…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00004671
CAS Number: 75-80-9
EINECS: 200-903-1
Merck Index: 12,09743

Order Now

Specification Test Results

Appearance White to yellow to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 73 to 82 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR