Toluene-3,4-dithiol 95%

Warning

Synonyms: 1,2-Dimercapto-4-methylbenzene4-Methyl-1,2-dimercaptobenzene4-Methylbenzene-1,2-dithiol3,4-Toluenedithiol

AC42118-0010 AC42118-0050
200032-978EA 142 USD
200032-978 200038-960
Toluene-3,4-dithiol 95%
Toluene-3,4-dithiol
Formula: C₇H₈S₂
MW: 156.27 g/mol
Boiling Pt: 185 °C (84 mmHg)
Melting Pt: 27 °C
Density: 1.18 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00004844
CAS Number: 496-74-2
EINECS: 207-828-3
UN: 2810
Merck Index: 12,09669

Order Now

Specification Test Results

Appearance White to yellow Low melting crystalline solid
Infrared spectrum Conforms
GC ≥94 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR