N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride 98+%

Warning

Synonyms: N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochlorideN1,N1,N4,N4-Tetramethylbenzene-1,4-diamine dihydrochlorideTMPD 2 HClN,N,N',N'-Tetramethyl-1,4-phenylenediammonium dichloride

AC42060-0050 AC42060-0250 AC42060-1000
200007-074EA 49.4 USD
200007-074 200013-502 102777-440
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
Formula: C₁₀H₁₈Cl₂N₂
MW: 237.17 g/mol
Melting Pt: 220…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00012482
CAS Number: 637-01-4
EINECS: 211-274-8
Merck Index: 12,09367
https://us.vwr.com/stibo/web/std.lang.all/61/62/2346162.jpg

Order Now

Specification Test Results

Appearance White to beige to grey to light brown Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 98.0 to 102.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR