Tetrahexylammonium bromide 99%

Warning

AC42040-1000
102777-040EA 268 USD
102777-040
Tetrahexylammonium bromide 99%
Tetrahexylammonium bromide
Formula: C₂₄H₅₂BrN
MW: 434.59 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011858
CAS Number: 4328-13-6
EINECS: 224-363-1

Order Now

Specification Test Results

Appearance White to light yellow Powder or flakes
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
GC ≥98.5 %
Color scale ≤15 APHA
Stabilizer 40 to 60 ppm MEHQ

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR