Tetrachlorocyclopropene 98%
|
Warning
|
AC42019-0050
AC42019-0250
200020-836EA
148
USD
200020-836
200044-762
Tetrachlorocyclopropene 98%
Tetrachlorocyclopropene
Formula:
C₃Cl₄ MW: 177.84 g/mol Boiling Pt: 125…130 °C Storage Temperature: Refrigerator |
CAS Number:
6262-42-6 EINECS: 228-418-0 ADR: 6.1,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5063 to 1.5083 (20 °C, 589 nm) |
GC | ≥97.5 % |
Stabilizer | Approx. 0.01 % Citric acid |
Relative density | (20/4°C) 1.026 to 1.036 |
Acid value | ≤5 mg KOH/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...