Sodium periodate 99.8+% ACS

Danger

Synonyms: Sodium metaperiodate

AC41961-1000 AC41961-5000
200007-698EA 534 USD
200007-698 102775-904
Sodium periodate 99.8+% ACS
Sodium periodate
Formula: NaIO₄
MW: 213.89 g/mol
Melting Pt: 300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 3085
ADR: 5.1,I
Merck Index: 12,08786

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Iodometric 99.8 to 100.3 % (on dried basis)
Other halogens (as Cl) ≤0.02 %
Manganese (Mn) ≤3 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR