Polyethylene glycol 1,450

AC41804-1000 AC41804-0030 AC41804-0010
200014-128EA 18.09 USD
200014-128 200020-452 200014-130
Polyethylene glycol 1,450
Polyethylene glycol 1,450
Formula: H(OCH₂CH₂)nOH
MW: 1450 g/mol
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR