N-Benzylaniline 99%

Synonyms: N-Phenylbenzylamine((Phenylmethyl)amino)benzene

AC41739-0050 AC41739-5000 AC41739-0250 AC41739-1000
200032-326EA 163 USD
200032-326 103466-190 200044-266 200044-264
N-Benzylaniline 99%
N-Benzylaniline
Formula: C₆H₅CH₂NHC₆H₅
MW: 183.25 g/mol
Boiling Pt: 306…307 °C (760 mmHg)
Melting Pt: 34…38 °C
Density: 1.06 g/cm³
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00003018
CAS Number: 103-32-2
EINECS: 203-100-4
Merck Index: 12,01161
https://us.vwr.com/stibo/web/std.lang.all/98/43/769843.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR