Pempidine 95%

Danger

AC41696-0050
200008-390EA 94.6 USD
200008-390
Pempidine 95%
Pempidine
Formula: C₁₀H₂₁N
MW: 155.28 g/mol
Boiling Pt: 187…188 °C
Melting Pt: 11 °C
Density: 0.858 g/cm³
Flash Pt: 50 °C
MDL Number: MFCD00006493
CAS Number: 79-55-0
EINECS: 201-211-2
UN: 1992
ADR: 3,III
Merck Index: 12,07207

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR