Pentafluoropropionic anhydride 98%
|
Danger
|
Synonyms:
Pentafluoropropanoic anhydride
, PFPA
, Perfluoropropanoic anhydride
AC41693-0250
AC41693-0050
AC41693-1000
200003-922EA
124
USD
200003-922
200013-926
103528-638
Pentafluoropropionic anhydride 98%
Pentafluoropropionic anhydride
Formula:
C₆F₁₀O₃ MW: 310.05 g/mol Boiling Pt: 69 °C Density: 1.57 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00000429 CAS Number: 356-42-3 EINECS: 206-604-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...