Sodium octyl sulfate 99% for HPLC

Danger

Synonyms: Octyl sulfate, sodium saltOctyl sulphuric acid saltOctyl sulphate, sodium saltOctyl sulfuric acid salt

AC41650-0250 AC41650-1000
103518-886EA 2242 USD
103518-886 200007-238
Sodium octyl sulfate 99% for HPLC
Sodium octyl sulphate
Formula: C₈H₁₇NaO₄S
MW: 232.28 g/mol
Melting Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00007470
CAS Number: 142-31-4
EINECS: 205-535-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR