Nitrilotriacetic acid 99%

Warning

Synonyms: 2,2',2''-Nitrilotriacetic acidNTA

AC41574-0010 AC41574-0050 AC41574-2500
200013-678EA 125 USD
200013-678 200007-176 200007-178
Nitrilotriacetic acid 99%
Nitrilotriacetic acid
Formula: C₆H₉NO₆
MW: 191.14 g/mol
Boiling Pt: 498 °C (1013 hPa)
Melting Pt: 245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004287
CAS Number: 139-13-9
EINECS: 205-355-7
ADR: 8,II
Merck Index: 12,06675

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR