Nickel (II) sulfate hexahydrate 98+% ACS

Danger

Synonyms: Nickelous sulphate, HexahydrateNickelous sulfate, HexahydrateNickel sulphate hexahydrateNickel sulfate hexahydrate

AC41561-0051 AC41561-5000 AC41561-0010
200003-902EA 117 USD
200003-902 103528-630 102776-480
Nickel (II) sulfate hexahydrate 98+% ACS
Nickel sulphate hexahydrate
Formula: NiSO₄·6H₂O
MW: 262.85 g/mol
Melting Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00149813
CAS Number: 10101-97-0
EINECS: 232-104-9
UN: 3077
ADR: 9,III
Merck Index: 13,06541

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR