Morpholine 99+% ACS

Danger

AC41516-0050 AC41516-0025
200007-054EA 17.7 USD
200007-054 102776-966
Morpholine 99+% ACS
Morpholine
Formula: C₄H₉NO
MW: 87.12 g/mol
Boiling Pt: 128.9 °C (1013 hPa)
Melting Pt: –5 °C
Density: 0.99 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00005972
CAS Number: 110-91-8
EINECS: 203-815-1
UN: 2054
ADR: 8,I
Merck Index: 12,06362
https://us.vwr.com/stibo/web/std.lang.all/82/95/2298295.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR