Methyl valerate 99%

Warning

Synonyms: Methyl PentanoateValeric acid methyl ester

AC41505-0250
200019-776EA 32.8 USD
200019-776
Methyl valerate 99%
Methyl valerate
Formula: C₆H₁₂O₂
Boiling Pt: 126 °C
Melting Pt: –91 °C
Density: 0.89 g/cm³
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00009478
CAS Number: 624-24-8
EINECS: 210-838-0
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR