Methyl-4-iodobenzoate 98%

Synonyms: 4-Iodobenzoic acid methyl esterMethyl 4-iodobenzoate

AC41446-1000 AC41446-0250
200007-004EA 112 USD
200007-004 200013-326
Methyl-4-iodobenzoate 98%
Methyl 4-iodobenzoate
Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00016353
CAS Number: 619-44-3
EINECS: 210-597-1
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR