Lithium stearate 95%

Synonyms: Stearic acid lithium saltLithium Stearate

AC41334-0250 AC41334-1000
200019-514EA 25.6 USD
200019-514 200052-750
Lithium stearate 95%
Lithium stearate
Formula: C₁₈H₃₅LiO₂
MW: 290.42 g/mol
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00042032
CAS Number: 4485-12-5
EINECS: 224-772-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR