Lithium dodecyl sulfate 99+%

Warning

Synonyms: Lauryl sulfate lithium saltLithium lauryl sulphateDodecyl sulfate lithium saltLithium lauryl sulfateDodecyl sulphate lithium saltLauryl sulphate lithium saltDodecyl sulphuric acid lithium saltDodecyl sulfuric acid lithium salt

AC41330-1000
103528-612EA 367 USD
103528-612
Lithium dodecyl sulfate 99+%
Lithium dodecyl sulphate
Formula: C₁₂H₂₅LiO₄S
MW: 272.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR