4-Isopropylbenzoic acid 98+%

Warning

AC41280-0050 AC41280-0250
200029-860EA 51.8 USD
200029-860 103512-484
4-Isopropylbenzoic acid 98+%
4-Isopropylbenzoic acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Melting Pt: 116…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00002564
CAS Number: 536-66-3
EINECS: 208-642-5
Merck Index: 12,02684

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR