4-Iodobenzonitrile 99%

Danger

Synonyms: p-Iodobenzonitrile

AC41238-0050 AC41238-0250
102775-932EA 104 USD
102775-932 200003-848
4-Iodobenzonitrile 99%
4-Iodobenzonitrile
Formula: C₇H₄IN
MW: 229.02 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00051310
CAS Number: 3058-39-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR