3'-Hydroxyacetophenone 99+%

Warning

AC41191-1000 AC41191-0050
200023-322EA 16.3 USD
200023-322 200017-092
3'-Hydroxyacetophenone 99+%
3'-Hydroxyacetophenone
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00002298
CAS Number: 121-71-1
EINECS: 204-494-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR