1-Heptanesulfonic acid sodium salt monohydrate for HPLC

Warning

Synonyms: Sodium 1-heptanesulphonate monohydrateSodium 1-heptanesulfonate monohydrate

AC41127-0250 AC41127-5000 AC41127-1000
103518-866EA 1549 USD
103518-866 200003-830 200004-734
1-Heptanesulfonic acid sodium salt monohydrate for HPLC
1-Heptanesulphonic acid sodium salt monohydrate
Formula: C₇H₁₅NaO₃S·1H₂O
MW: 220.27 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149550
CAS Number: 207300-90-1
EINECS: 245-210-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR