Guanidinium thiocyanate 99% for biochemistry

Danger

Synonyms: Guanidinium rhodanide Guanidinium thiocyanate

AC41111-0010 AC41111-1000 AC41111-2500
102777-454EA 547 USD
102777-454 200012-624 200012-622
Guanidinium thiocyanate 99% for biochemistry
Guanidinium thiocyanate
Formula: NH₂C(=NH)NH₂·HSCN
MW: 118.16 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR