Glycolic acid 70%

Danger

Synonyms: 2-Hydroxy acid2-Hydroxyacetic acid

AC41103-0250 AC41103-0010 AC41103-2500
103528-596EA 271 USD
103528-596 200057-706 200057-708
Glycolic acid 70%
Glycolic acid
Formula: C₂H₄O₃
MW: 76.05 g/mol
Melting Pt: 10 °C
Density: 1.27 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS Number: 79-14-1
EINECS: 201-180-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR