Gallic acid 98%

Warning

Synonyms: 3,4,5-Trihydroxybenzoic acid

AC41086-0010 AC41086-0051 AC41086-0050 AC41086-2500
103512-432EA 1308 USD
103512-432 200012-554 200012-556 200031-444
Gallic acid 98%
Gallic acid
Formula: C₇H₆O₅
MW: 170.12 g/mol
Melting Pt: 253 °C (dec.)
Density: 1.694 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00002510
CAS Number: 149-91-7
EINECS: 205-749-9
Merck Index: 13,04366

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR