Hexafluorosilicic acid 25% (w/w) in aqueous solution
|
Danger
|
Synonyms:
Fluosilicic acid
AC41071-0025
AC41071-5000
200057-604EA
45.69
USD
200057-604
103533-104
Hexafluorosilicic acid 25% (w/w) in aqueous solution
Hexafluorosilicic acid
Formula:
H₂F₆Si MW: 144.09 Density: 1.27 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00036289 EINECS: 241-034-8 UN: 1778 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...