Ethyl valerate 99%

Warning

Synonyms: Valeric acid ethyl esterEthyl Pentanoate

AC41047-0050 AC41047-5000 AC41047-1000
200043-542EA 26 USD
200043-542 200012-456 102774-214
Ethyl valerate 99%
Ethyl valerate
Formula: C₇H₁₄O₂
Boiling Pt: 145 °C
Melting Pt: –91 °C
Density: 0.87 g/cm³
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00009479
CAS Number: 539-82-2
EINECS: 208-726-1
UN: 3272
ADR: 3,III
Merck Index: 12,10042

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR