Ethyl propenyl ether cis- and trans-mixture 98%

Danger

Synonyms: Ethyl 1-Propenyl ether

AC41037-0250 AC41037-1000 AC41037-0100
200012-440EA 258 USD
200012-440 200012-442 102774-688
Ethyl propenyl ether cis- and trans-mixture 98%
Ethyl propenyl ether
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 55…65 °C (1013 hPa)
Density: 0.778 g/cm³
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00015184
CAS Number: 928-55-2
EINECS: 213-176-0
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR