4-Amino-N,N-dimethylaniline 97%

Danger

Synonyms: N,N-Dimethylbenzene-p-diamineN,N-Dimethylbenzene-1,4-diamineN,N-Dimethyl-1,4-phenylenediamineN,N-Dimethyl-p-phenylenediamine

AC40846-5000 AC40846-1000 AC40846-0250 AC40846-0050
102777-356EA 482 USD
102777-356 200007-108 102774-702 102775-816
4-Amino-N,N-dimethylaniline 97%
4-Amino-N,N-dimethylaniline
Formula: C₈H₁₂N₂
MW: 136.2 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 35 °C
Density: 1.09 g/cm³
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00007860
CAS Number: 99-98-9
EINECS: 202-807-5
UN: 2811
ADR: 6.1,II
Merck Index: 13,33280

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR