Dimethyl glyoxime 99+% ACS

Danger

Synonyms: Diacetyl dioxime2,3-Butanedione dioximeDiacetyl glyoxime

AC40833-1000 AC40833-0025 AC40833-0250 AC40833-5000
102777-700EA 826 USD
102777-700 200018-842 200018-844 200012-118
Dimethyl glyoxime 99+% ACS
Dimethyl glyoxime
Formula: CH₃C(=NOH)C(=NOH)CH₃
MW: 116.12 g/mol
Boiling Pt: 53 °C (8 mmHg)
Melting Pt: 239…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00002117
CAS Number: 95-45-4
EINECS: 202-420-1
UN: 2926
ADR: 4.1,III
Merck Index: 13,03272

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR