N,N-Dimethylcyclohexylamine 99%

Danger

Synonyms: N-CyclohexyldimethylamineDimethylaminocyclohexane

AC40825-5000 AC40825-0025
200026-046EA 56 USD
200026-046 102776-208
N,N-Dimethylcyclohexylamine 99%
N,N-Dimethylcyclohexylamine
Formula: C₈H₁₇N
MW: 127.23 g/mol
Boiling Pt: 160…161 °C (1013 hPa)
Melting Pt: –60 °C
Density: 0.84 g/cm³
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00003844
CAS Number: 98-94-2
EINECS: 202-715-5
UN: 2264
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR