Dihexylamine 99+%

Danger

AC40759-5000 AC40759-1000
103528-578EA 44.5 USD
103528-578 102775-890
Dihexylamine 99+%
Dihexylamine
Formula: C₁₂H₂₇N
MW: 185.35 g/mol
Boiling Pt: 236 °C (760 mmHg)
Melting Pt: 3 °C
Density: 0.8 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00009521
CAS Number: 143-16-8
EINECS: 205-588-4
UN: 2927
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR