N,N-Dibutylaniline ≥99%

Warning

AC40657-0250
200026-022EA 53.5 USD
200026-022
N,N-Dibutylaniline ≥99%
N,N-Dibutylaniline
Formula: (C₆H₅)N(CH₂CH₂CH₂CH₃)₂
MW: 205.34 g/mol
Boiling Pt: 265…275 °C (1013 hPa)
Melting Pt: –32 °C
Density: 0.90368 g/cm³ (20 °C)
MDL Number: MFCD00035799
CAS Number: 613-29-6
EINECS: 210-335-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR