1,3-Dibromoacetone 70% tech.

Warning

Synonyms: 1,3-Dibromopropan-2-one

AC40655-0250
200008-460EA 402 USD
200008-460
1,3-Dibromoacetone 70% tech.
1,3-Dibromoacetone
Formula: BrH₂CC(O)CH₂Br
MW: 215.87 g/mol
Boiling Pt: 95 °C (20 mmHg)
Melting Pt: 28 °C
MDL Number: MFCD00013540
CAS Number: 816-39-7
EINECS: 212-430-8
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR