2,4-Dibromophenol 99%

Danger

AC40651-0050 AC40651-1000
200016-080EA 128 USD
200016-080 200022-170
2,4-Dibromophenol 99%
2,4-Dibromophenol
Formula: C₆H₄Br₂O
MW: 251.91 g/mol
Boiling Pt: 154 °C (11 mmHg)
Melting Pt: 36…39 °C
Flash Pt: >110 °C
MDL Number: MFCD00002149
CAS Number: 615-58-7
EINECS: 210-436-5
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR