5-Chlorosalicylaldehyde 98%

Warning

Synonyms: 5-Chloro-2-hydroxybenzaldehyde

AC40503-0050 AC40503-1000 AC40503-0250
200042-276EA 290 USD
200042-276 200017-822 102773-988
5-Chlorosalicylaldehyde 98%
5-Chlorosalicylaldehyde
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 105 °C (12 mmHg)
Melting Pt: 102…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00003331
CAS Number: 635-93-8
EINECS: 211-244-4
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR