4-Chlorophenylhydrazinium chloride 97%

Warning

Synonyms: 1-(4-Chlorophenyl)hydrazine hydrochloride4-Chlorophenylhydrazine hydrochloride

AC40495-1000 AC40495-0050 AC40495-0250
103514-754EA 270 USD
103514-754 200013-908 200029-756
4-Chlorophenylhydrazinium chloride 97%
4-Chlorophenylhydrazinium chloride
Formula: C₆H₈Cl₂N₂
MW: 179.05 g/mol
Melting Pt: 203 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012943
CAS Number: 1073-70-7
EINECS: 214-030-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR