2-Chloro-5-nitrobenzaldehyde 99%

Warning

AC40487-1000 AC40487-0250
200051-830EA 179 USD
200051-830 200063-922
2-Chloro-5-nitrobenzaldehyde 99%
2-Chloro-5-nitrobenzaldehyde
Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Melting Pt: 76…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00007293
CAS Number: 6361-21-3
EINECS: 228-830-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR