1-Chlorohexane ≥95%

Supplier: ACROS ORGANICS MS

Warning

AC40464-5000 AC40464-0050 AC40464-1000
102776-058EA 126.37 USD
102776-058 200040-232 200015-764
1-Chlorohexane ≥95%
1-Chlorohexane
Formula: C₆H₁₃Cl
MW: 120.62 g/mol
Boiling Pt: ∼135 °C (1013 hPa)
Melting Pt: –94 °C
Density: 0.872 g/cm³ (20 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00001018
CAS Number: 544-10-5
EINECS: 208-859-5
UN: 1993
ADR: 3,III
Merck Index: 13,02163

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4210 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR